Learn More
Cole-Parmer TPE Tubing, Clear, 5/16" ID x 7/16" OD; 50 Ft
Supplier:  Cole-Parmer 5011613
Encompass_Preferred
The Cole-Parmer Thermoplastic Elastomer (TPE) tubing is soft and flexible tubing that is durable enough to provide a long service life. This tubing is designed for use with peristaltic pumps. This TPE tubing does not contain PVC, DEHP, phthalates, or plasticizers, making them ideal for various pharmaceutical, medical, food, and beverage applications.
Catalog No. 50-255-7919