Learn More
TARGETMOL CHEMICALS INC TPEN 200MG
SDP
Catalog No. 502878640
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TPEN (TPEDA) is a specific cell-permeable heavy metal chelator. Purity 99.79%
Catalog No. 50-287-8640 Supplier TARGETMOL CHEMICALS INC Supplier No. T3952200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TPEN (TPEDA) is a specific cell-permeable heavy metal chelator. Purity 99.79%