Learn More
Selleck Chemical LLC TPEN-25mg
SDP
Catalog No. 502389641
Encompass_Preferred
TPEN (TPEDA) is a specific cell-permeable heavy metal chelator that induces apoptosis in NB4 cells via negatively regulating intracellular NO and Zn2+ signaling. TPEN (TPEDA) may be a potential therapeutic strategy for APL (acute promyelocytic leukemia).TPEN induces DNA damage and increases intracellular ROS production and also inhibits cell proliferation.
Catalog No. 50-238-9641 Supplier Selleck Chemical LLC Supplier No. S696225MG
TPEN (TPEDA) is a specific cell-permeable heavy metal chelator that induces apoptosis in NB4 cells via negatively regulating intracellular NO and Zn2+ signaling. TPEN (TPEDA) may be a potential therapeutic strategy for APL (acute promyelocytic leukemia).TPEN induces DNA damage and increases intracellular ROS production and also inhibits cell proliferation.