Learn More
Medchemexpress LLC HY-100202 50mg Medchemexpress, TPEN CAS:16858-02-9 Purity:>98%
SDP
Catalog No. 501931749
Encompass_Preferred
Medchemexpress, HY-100202 50mg TPEN CAS:16858-02-9 TPEN is a specific cell-permeable heavy metal chelator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1749 Supplier Medchemexpress LLC Supplier No. HY10020250MG
Medchemexpress, HY-100202 50mg TPEN CAS:16858-02-9 TPEN is a specific cell-permeable heavy metal chelator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.