Learn More
AdipoGen TPEN
SDP
Catalog No. 502560034
Encompass_Preferred
Chemical. CAS 16858-02-9. Formula C26H28N6. MW 424.5. TPEN is a cell permeable metal ion chelator. TPEN is a chelator that has a high affinity for zinc Zn2, next to other transition metals Zn2 >Fe2 >Mn2 and is used in zinc treatment studies to limit intra- and extracellular concentrations of the mineral. TPEN has lower affinity for Ca2 and Mg2 and can also form complexes with other soft metal ions such as Cd2. In addition to a heavy metal chelator, TPEN is also known to be an apoptosis inducer. Depletion of zinc by TPEN induces apoptosis in several cell lines. TPEN shows anticancer activity through induction of caspase-dependent apoptosis, via reactive oxygen species ROS signaling mechanism and inhibiting cell autophagy. TPEN also inhibits RNA binding protein Lin28 and has been shown to be useful in extracting Abeta depostis from Alzheimers disease brain tissue.
Catalog No. 50-256-0034 Supplier AdipoGen Supplier No. CDXT0044G001
Chemical. CAS 16858-02-9. Formula C26H28N6. MW 424.5. TPEN is a cell permeable metal ion chelator. TPEN is a chelator that has a high affinity for zinc Zn2, next to other transition metals Zn2 >Fe2 >Mn2 and is used in zinc treatment studies to limit intra- and extracellular concentrations of the mineral. TPEN has lower affinity for Ca2 and Mg2 and can also form complexes with other soft metal ions such as Cd2. In addition to a heavy metal chelator, TPEN is also known to be an apoptosis inducer. Depletion of zinc by TPEN induces apoptosis in several cell lines. TPEN shows anticancer activity through induction of caspase-dependent apoptosis, via reactive oxygen species ROS signaling mechanism and inhibiting cell autophagy. TPEN also inhibits RNA binding protein Lin28 and has been shown to be useful in extracting Abeta depostis from Alzheimers disease brain tissue.