Learn More
Abcam TPEN, Metal ion chelator, 100MG
Catalog No. 502706896
Encompass_Preferred
MW 424.5 Da, Purity >99%. Metal ion chelator. Decreases intracellular zinc levels. Induces apoptosis. Causes defects in oocyte maturation. Modulates hippocampal neurogenesis following seizure. Cell permeable. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6896 Supplier Abcam Supplier No. AB141111100MG
MW 424.5 Da, Purity >99%. Metal ion chelator. Decreases intracellular zinc levels. Induces apoptosis. Causes defects in oocyte maturation. Modulates hippocampal neurogenesis following seizure. Cell permeable. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement