Learn More
AdipoGen TPh A (5 mg)
SDP
Supplier:  AdipoGen AGCR13507M005
Encompass_Preferred
Chemical. CAS: 21306-65-0. Formula: C21H21NO3S2. MW: 399.5. Nuclear protein pirin inhibitor. Inhibits the interaction between Bcl3 oncoprotein and the nuclear protein pirin. Melanoma cell migration inhibitor.
Catalog No. 50-205-3885