Learn More
Abcam TPh A, nuclear protein pirin inhibitor, 5MG
Catalog No. 502705832
Encompass_Preferred
MW 399.5 Da, Purity >98%. Inhibitor of nuclear protein pirin. Inhibits the interaction between Bcl3 oncoprotein and pirin. Melanoma cell migration inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5832 Supplier Abcam Supplier No. AB1443765MG
MW 399.5 Da, Purity >98%. Inhibitor of nuclear protein pirin. Inhibits the interaction between Bcl3 oncoprotein and pirin. Melanoma cell migration inhibitor.
The product is subject to the following: Abcam Restricted Use Statement