Learn More
Medchemexpress LLC TPh A (Triphenyl Compound A) | 21306-65-0 | 99.8% | C21H21NO3S2 | 50 MG
SDP
Catalog No. 5000353922
Encompass_Preferred
TPh A is a potent inhibitor of the nuclear protein pirin, binding specifically with a Ki of 0.6 μM. It disrupts the formation of the bcl3-pirin complex and serves as a small molecule tool to regulate pirin in cells. This product is soluble in DMSO at 250 mg/mL.
  • Potent inhibitor of nuclear protein pirin
  • Binds specifically to pirin with a Ki of 0.6 μM
  • Disrupts bcl3-pirin complex formation
  • Regulates pirin in cells
  • Exhibits IC50 values of 27 μM, 20 μM, and 26 μM for PC3, HL60, and HT29 cells
  • Inhibits cell migration in WM266-4 and SK-MEL-28 cells
Catalog No. 50-003-53922 Supplier Medchemexpress LLC Supplier No. HY1445450MG
TPh A is a potent inhibitor of the nuclear protein pirin, binding specifically with a Ki of 0.6 μM. It disrupts the formation of the bcl3-pirin complex and serves as a small molecule tool to regulate pirin in cells. This product is soluble in DMSO at 250 mg/mL.
  • Potent inhibitor of nuclear protein pirin
  • Binds specifically to pirin with a Ki of 0.6 μM
  • Disrupts bcl3-pirin complex formation
  • Regulates pirin in cells
  • Exhibits IC50 values of 27 μM, 20 μM, and 26 μM for PC3, HL60, and HT29 cells
  • Inhibits cell migration in WM266-4 and SK-MEL-28 cells