Learn More
Medchemexpress LLC TPh A (Triphenyl compound A) | 21306-65-0 | MFCD01312191 | 99.8% | C21H21NO3S2 | 10MG
SDP
Catalog No. 5000261849
Encompass_Preferred
TPh A (Triphenyl Compound A) is a small-molecule inhibitor of the nuclear protein pirin that binds with a Ki of about 0.6 μM and disrupts BCL3-pirin complex formation. Supplied as a white to off-white solid for research use, it has a reported molecular weight of 399.53 Da and high analytical purity.
  • Potent pirin inhibitor with a Ki of 0.6 μM.
  • Disrupts formation of the BCL3-pirin complex.
  • High reported purity suitable for biochemical and cell-based assays.
  • Available in solid and solution formats for flexible workflow integration.
  • Soluble in common organic solvents such as DMSO for stock preparation.
  • Stable as a powder under low-temperature storage for long-term use.
Catalog No. 50-002-61849 Supplier Medchemexpress LLC Supplier No. HY1445410MG
TPh A (Triphenyl Compound A) is a small-molecule inhibitor of the nuclear protein pirin that binds with a Ki of about 0.6 μM and disrupts BCL3-pirin complex formation. Supplied as a white to off-white solid for research use, it has a reported molecular weight of 399.53 Da and high analytical purity.
  • Potent pirin inhibitor with a Ki of 0.6 μM.
  • Disrupts formation of the BCL3-pirin complex.
  • High reported purity suitable for biochemical and cell-based assays.
  • Available in solid and solution formats for flexible workflow integration.
  • Soluble in common organic solvents such as DMSO for stock preparation.
  • Stable as a powder under low-temperature storage for long-term use.