Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPH in Soil - Low - PT, MilliporeSigma™ Supelco™

Catalog No. 111016934 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6934 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6934 Supplier MilliporeSigma Supelco Supplier No. SPE026TXL30G

missing translation for 'validMainframeMsg'

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 109-66-0
Flash Point -49°C - closed cup - Solvent
Type Proficiency Testing Material
Quantity 30 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.