Learn More
ABclonal Technology TPH2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A145201000UL
Encompass_Preferred
This gene encodes a member of the pterin-dependent aromatic acid hydroxylase family. The encoded protein catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
Catalog No. 50-001-06964