Learn More
Medchemexpress LLC HY-100463 10mg Medchemexpress, TPI-1 CAS:79756-69-7 Purity:>98%
SDP
Catalog No. 502028621
Encompass_Preferred
Medchemexpress, HY-100463 10mg TPI-1 CAS:79756-69-7 TPI-1, also known as Tyrosine Phosphatase Inhibitor 1, is a SHP-1 inhibitor; inhibits recombinant SHP-1 with an IC50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8621 Supplier Medchemexpress LLC Supplier No. HY10046310MG
Medchemexpress, HY-100463 10mg TPI-1 CAS:79756-69-7 TPI-1, also known as Tyrosine Phosphatase Inhibitor 1, is a SHP-1 inhibitor; inhibits recombinant SHP-1 with an IC50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.