Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TPI-287 | 849213-15-6 | 99.5% | 1 MG
SDP

Catalog No. 5000341543
Encompass_Preferred

TPI-287 is a blood-brain barrier-permeable microtubule stabilizer that can significantly reduce metastatic colonization of breast cancer in the brain. It has been shown to effectively reduce tumor volume and inhibit tumor growth in mouse tumor models.

  • Blood-brain barrier permeable microtubule stabilizer
  • Significantly reduces metastatic colonization of breast cancer in the brain
  • Effectively reduces tumor volume and inhibits tumor growth
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month, under nitrogen
  • Soluble in DMSO: ≥ 100 mg/mL

Catalog No. 50-003-41543 Supplier Medchemexpress LLC Supplier No. HY1268991MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TPI-287 is a blood-brain barrier-permeable microtubule stabilizer that can significantly reduce metastatic colonization of breast cancer in the brain. It has been shown to effectively reduce tumor volume and inhibit tumor growth in mouse tumor models.

  • Blood-brain barrier permeable microtubule stabilizer
  • Significantly reduces metastatic colonization of breast cancer in the brain
  • Effectively reduces tumor volume and inhibits tumor growth
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month, under nitrogen
  • Soluble in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.