Learn More
Medchemexpress LLC TPI-287 | 849213-15-6 | 99.5% | 10 MG
SDP
Catalog No. 5000341331
Encompass_Preferred
TPI-287 is a blood-brain barrier-permeable microtubule stabilizer that can significantly reduce metastatic colonization of breast cancer in the brain. It has shown to effectively decrease tumor volume and inhibit tumor growth in mouse models.
  • Blood-brain barrier permeable microtubule stabilizer
  • Reduces metastatic colonization of breast cancer in the brain
  • Decreases tumor volume and inhibits tumor growth in mouse models
Catalog No. 50-003-41331 Supplier Medchemexpress LLC Supplier No. HY12689910MG
TPI-287 is a blood-brain barrier-permeable microtubule stabilizer that can significantly reduce metastatic colonization of breast cancer in the brain. It has shown to effectively decrease tumor volume and inhibit tumor growth in mouse models.
  • Blood-brain barrier permeable microtubule stabilizer
  • Reduces metastatic colonization of breast cancer in the brain
  • Decreases tumor volume and inhibits tumor growth in mouse models