Learn More
Medchemexpress LLC TPI-287 | 849213-15-6 | 99.5% | 25 MG
SDP
Catalog No. 5000341397
Encompass_Preferred
TPI-287 is a blood-brain barrier-permeable microtubule stabilizer that can significantly reduce metastatic colonization of breast cancer in the brain. This product is for research use only.
  • Blood-brain barrier-permeable microtubule stabilizer
  • Significantly reduces metastatic colonization of breast cancer in the brain
  • For research use only
Catalog No. 50-003-41397 Supplier Medchemexpress LLC Supplier No. 25MGHY126899
TPI-287 is a blood-brain barrier-permeable microtubule stabilizer that can significantly reduce metastatic colonization of breast cancer in the brain. This product is for research use only.
  • Blood-brain barrier-permeable microtubule stabilizer
  • Significantly reduces metastatic colonization of breast cancer in the brain
  • For research use only