Learn More
Medchemexpress LLC TPI-287 | 849213-15-6 | 99.5% | 50 MG
SDP
Catalog No. 5000342193
Encompass_Preferred
TPI-287 is a blood-brain barrier-permeable microtubule stabilizer.
  • Can significantly reduce metastatic colonization of breast cancer in the brain.
Catalog No. 50-003-42193 Supplier Medchemexpress LLC Supplier No. HY12689950MG
TPI-287 is a blood-brain barrier-permeable microtubule stabilizer.
  • Can significantly reduce metastatic colonization of breast cancer in the brain.