Learn More
Medchemexpress LLC Tpi-287 (taxane analogue) | 849213-15-6 | MFCD35060996 | 99.5% | 870.0 g/mol | C46H63NO15 | 5 MG
SDP
Catalog No. 5000264636
Encompass_Preferred
TPI-287 is a brain-penetrant taxane-class microtubule stabilizer intended for preclinical oncology research. It stabilizes microtubules, shows antineoplastic activity in models of metastatic breast cancer to the brain, and is supplied as a research-grade small molecule for laboratory investigation.
  • Blood-brain barrier-permeable microtubule stabilizer.
  • Demonstrated reduction of metastatic colonization in brain in preclinical studies.
  • High purity (~99.5% by HPLC) for reproducible experimental results.
  • Available in small-mass pack sizes suitable for in vitro and in vivo work.
  • Chemical formula C46H63NO15; molecular weight 870.0 g/mol.
Catalog No. 50-002-64636 Supplier Medchemexpress LLC Supplier No. HY1268995MG
TPI-287 is a brain-penetrant taxane-class microtubule stabilizer intended for preclinical oncology research. It stabilizes microtubules, shows antineoplastic activity in models of metastatic breast cancer to the brain, and is supplied as a research-grade small molecule for laboratory investigation.
  • Blood-brain barrier-permeable microtubule stabilizer.
  • Demonstrated reduction of metastatic colonization in brain in preclinical studies.
  • High purity (~99.5% by HPLC) for reproducible experimental results.
  • Available in small-mass pack sizes suitable for in vitro and in vivo work.
  • Chemical formula C46H63NO15; molecular weight 870.0 g/mol.