Learn More
MSC TPI Corporation Exhaust Fan, 18 in. Aluminum Blade, 1/8 HP, 3-Speed, 120 V, 1 ea.
Catalog No. 5000507516
Encompass
This 18 in. direct drive exhaust fan is engineered for effective air circulation in commercial environments. It features a totally enclosed air-over (TEAO) motor, providing reliable performance and thermal protection. The unit is designed for direct wall mounting, offering versatile installation options with or without an external shutter.
  • Features a 1/8 hp, 120V motor with three-speed operation
  • Includes an aluminum 3-paddle blade system for efficient airflow up to 2,300 cfm
  • Equipped with a pull chain switch for convenient speed control
  • Constructed with a powder-coated steel frame and wire guards for durability
  • Designed for easy direct wiring into a junction box
  • Manufactured in the U.S.A. with domestic and foreign components
Catalog No. 50-005-07516 Supplier MSC Supplier No. 84671726
This 18 in. direct drive exhaust fan is engineered for effective air circulation in commercial environments. It features a totally enclosed air-over (TEAO) motor, providing reliable performance and thermal protection. The unit is designed for direct wall mounting, offering versatile installation options with or without an external shutter.
  • Features a 1/8 hp, 120V motor with three-speed operation
  • Includes an aluminum 3-paddle blade system for efficient airflow up to 2,300 cfm
  • Equipped with a pull chain switch for convenient speed control
  • Constructed with a powder-coated steel frame and wire guards for durability
  • Designed for easy direct wiring into a junction box
  • Manufactured in the U.S.A. with domestic and foreign components