Learn More
MSC TPI Electric Baseboard Heater, 60 in, 240/208 V, 1250/938 W
Catalog No. 5000504206
Encompass
This electric baseboard heater from the 2900S Series provides efficient and safe heating. It features a durable steel front, stainless steel heating element, and aluminum fins, designed for easy wall mounting.
  • Stainless steel heating element for durability.
  • Aluminum fins for efficient heat distribution.
  • Durable painted 22 gauge steel front.
  • 20 gauge junction boxes with quick connect cable clamps and 6″ ground wires.
  • Over-temperature thermal limit for safety.
  • Full length wireway for organized wiring.
  • Easy wall mounting using keyhole knockouts.
  • Optional in-built thermostats can be field installed.
  • ETL-CUS certified for safety and quality.
  • Made in U.S.A.
Catalog No. 50-005-04206 Supplier MSC Supplier No. 00220921
This electric baseboard heater from the 2900S Series provides efficient and safe heating. It features a durable steel front, stainless steel heating element, and aluminum fins, designed for easy wall mounting.
  • Stainless steel heating element for durability.
  • Aluminum fins for efficient heat distribution.
  • Durable painted 22 gauge steel front.
  • 20 gauge junction boxes with quick connect cable clamps and 6″ ground wires.
  • Over-temperature thermal limit for safety.
  • Full length wireway for organized wiring.
  • Easy wall mounting using keyhole knockouts.
  • Optional in-built thermostats can be field installed.
  • ETL-CUS certified for safety and quality.
  • Made in U.S.A.