Learn More
MSC TPI Electric Baseboard Heater, 750W, 120V, 40 in, White
Catalog No. 5000503512
Encompass
This TPI Electric Baseboard Heater provides efficient heating for various applications. It is designed with robust materials and features for reliable performance and easy installation.
  • Available in white or ivory finishes.
  • Features a stainless steel heating element and aluminum fins.
  • Constructed with a painted 22 gauge steel front.
  • Includes 20 gauge junction boxes located at each end of the heater, equipped with quick connect cable clamps and 6-inch ground lead wires.
  • An over-temperature thermal limit extends the entire length of the heated area.
  • A full-length wireway is provided along the back of the heater; enclosed conduit is available as an option.
  • Can be wall-mounted using keyhole knockouts spaced every 4 inches at the back of the heater.
  • Optional in-built thermostats can be field-installed in the junction box.
Catalog No. 50-005-03512 Supplier MSC Supplier No. 00220848
This TPI Electric Baseboard Heater provides efficient heating for various applications. It is designed with robust materials and features for reliable performance and easy installation.
  • Available in white or ivory finishes.
  • Features a stainless steel heating element and aluminum fins.
  • Constructed with a painted 22 gauge steel front.
  • Includes 20 gauge junction boxes located at each end of the heater, equipped with quick connect cable clamps and 6-inch ground lead wires.
  • An over-temperature thermal limit extends the entire length of the heated area.
  • A full-length wireway is provided along the back of the heater; enclosed conduit is available as an option.
  • Can be wall-mounted using keyhole knockouts spaced every 4 inches at the back of the heater.
  • Optional in-built thermostats can be field-installed in the junction box.