Learn More
MSC TPI Electric Baseboard Heater, 84 in. Length, 240/208 Volt, 1750/1313 Watt
Catalog No. 5000504208
Encompass
This electric baseboard heater provides reliable warmth for various spaces. It features a durable stainless steel heating element and efficient aluminum fins. Designed for wall mounting, this unit includes a thermostat for easy temperature control and meets ETLus safety standards.
  • Made in the U.S.A.
  • Painted 22 gauge steel front.
  • 20 gauge junction boxes on each end with quick connect cable clamps and 6 in. ground lead wires.
  • Over-temperature thermal limit extends the entire length of the heated area.
  • Full length wireway provided along the back of the heater.
  • Mounts to the wall using key hole knockouts every 4 in. at the back.
  • Stainless steel heating element.
  • Aluminum fins.
  • Thermostat included.
  • Designed for wall mount installation.
  • Meets ETLus standards.
Catalog No. 50-005-04208 Supplier MSC Supplier No. 00220947
This electric baseboard heater provides reliable warmth for various spaces. It features a durable stainless steel heating element and efficient aluminum fins. Designed for wall mounting, this unit includes a thermostat for easy temperature control and meets ETLus safety standards.
  • Made in the U.S.A.
  • Painted 22 gauge steel front.
  • 20 gauge junction boxes on each end with quick connect cable clamps and 6 in. ground lead wires.
  • Over-temperature thermal limit extends the entire length of the heated area.
  • Full length wireway provided along the back of the heater.
  • Mounts to the wall using key hole knockouts every 4 in. at the back.
  • Stainless steel heating element.
  • Aluminum fins.
  • Thermostat included.
  • Designed for wall mount installation.
  • Meets ETLus standards.