Learn More
MSC TPI Fan Forced Unit Heater, 20.0 kW, 480V, 1 ea
Catalog No. 5000504464
Encompass
The TPI 5100 Series electric unit heater provides efficient heating with a 20.0 kW output, operating on 480V, 3-phase power, delivering 68.3 Btu/h. It is designed for flexible installation with wall or ceiling mounting options.
  • Heavy 18-gauge welded steel cabinet with powder-coated finish
  • Hinged and latched access door for simplified maintenance
  • Individually adjustable louvers for directional airflow control up to 15°
  • Copper-clad steel sheath heating elements with continuously brazed steel fins
  • Automatic reset limit controls for over-temperature protection
  • Thermally protected, permanently lubricated fan motor
  • Fan override purges residual heat from the unit at shutdown
  • Horizontal air throw of 32 ft. and 1100 CFM
Catalog No. 50-005-04464 Supplier MSC Supplier No. 01382522
The TPI 5100 Series electric unit heater provides efficient heating with a 20.0 kW output, operating on 480V, 3-phase power, delivering 68.3 Btu/h. It is designed for flexible installation with wall or ceiling mounting options.
  • Heavy 18-gauge welded steel cabinet with powder-coated finish
  • Hinged and latched access door for simplified maintenance
  • Individually adjustable louvers for directional airflow control up to 15°
  • Copper-clad steel sheath heating elements with continuously brazed steel fins
  • Automatic reset limit controls for over-temperature protection
  • Thermally protected, permanently lubricated fan motor
  • Fan override purges residual heat from the unit at shutdown
  • Horizontal air throw of 32 ft. and 1100 CFM