Learn More
MSC TPI Fan Forced Wall Heater, 240/208V, 4000/3000W, 1 ea
Catalog No. 5000506757
Encompass
The 3310 Series Fan Forced Wall Heater provides efficient heating for residential, commercial, and industrial indoor environments. This unit features heavy-gauge steel construction and a fan-forced design to deliver consistent warmth with an automatic reset thermal limit.
  • Powder coated 18 gauge steel front
  • Steel block fin element
  • Vane axial fan blade delivers 175 CFM
  • Dual wattage models are factory-wired to highest wattage
  • Built-in single pole thermostat with 0-110 deg F temperature range
  • Listed as cETLus for safety standards
  • Made in USA
Catalog No. 50-005-06757 Supplier MSC Supplier No. 76934165
The 3310 Series Fan Forced Wall Heater provides efficient heating for residential, commercial, and industrial indoor environments. This unit features heavy-gauge steel construction and a fan-forced design to deliver consistent warmth with an automatic reset thermal limit.
  • Powder coated 18 gauge steel front
  • Steel block fin element
  • Vane axial fan blade delivers 175 CFM
  • Dual wattage models are factory-wired to highest wattage
  • Built-in single pole thermostat with 0-110 deg F temperature range
  • Listed as cETLus for safety standards
  • Made in USA