This portable electric salamander heater provides clean, quiet, and odor-free fan-forced heat, designed for indoor use. It effectively circulates room air without adding moisture, featuring a robust design for dependable heating on various job sites.
- Equipped with 15 kW power, 480 V, and 3-phase operation, delivering 51,195 BTU.
- Features six stainless steel finned heating elements for long life.
- Includes an automatic thermostat control with a range of 40–100°F and a fan-only option.
- Safety yellow enclosure incorporates front and rear safety screens and a high-limit thermal cutout switch with automatic reset.
- Mounted on a two-wheeled cart for enhanced portability and ease of movement.
- Utilizes an 800 CFM fan for efficient air circulation.