Learn More
MSC TPI Infrared Heater, 4,947 Btu, 120V, 1450W
Catalog No. 5000508214
Encompass
This infrared heater provides 4,947 Btu of heating power. Operating at 120V and 1450W, it draws 12.1 Amps, making it suitable for various heating applications.
  • Provides 4,947 Btu heating capacity
  • Operates on 120V
  • Delivers 1450W of power
  • Draws 12.1 Amps
  • Single phase operation
Catalog No. 50-005-08214 Supplier MSC Supplier No. 55620132
This infrared heater provides 4,947 Btu of heating power. Operating at 120V and 1450W, it draws 12.1 Amps, making it suitable for various heating applications.
  • Provides 4,947 Btu heating capacity
  • Operates on 120V
  • Delivers 1450W of power
  • Draws 12.1 Amps
  • Single phase operation