Learn More
MSC TPI Infrared Suspended Heater, 240 Volt, 3200 Watt, 1 ea
Catalog No. 5000506955
Encompass
This infrared suspended heater operates at 240 Volt with a wattage of 3200 W, featuring 2 elements for efficient heating. It is suitable for both indoor and outdoor applications.
  • Two elements
  • One phase, single
  • Max BTU 10,922
  • Wattage of 3200 W
  • Overall length 24 in
  • Overall width 15 in
  • Overall height 10-7/8 in
  • Brown housing color
  • Suitable for indoor and outdoor use
  • Country of origin: UNITED STATES
Catalog No. 50-005-06955 Supplier MSC Supplier No. 76933365
This infrared suspended heater operates at 240 Volt with a wattage of 3200 W, featuring 2 elements for efficient heating. It is suitable for both indoor and outdoor applications.
  • Two elements
  • One phase, single
  • Max BTU 10,922
  • Wattage of 3200 W
  • Overall length 24 in
  • Overall width 15 in
  • Overall height 10-7/8 in
  • Brown housing color
  • Suitable for indoor and outdoor use
  • Country of origin: UNITED STATES