Learn More
MSC TPI Portable IR Flat Panel Heater, Roll-About, 240V, 14,972 BTU
Catalog No. 5000509073
Encompass
This TPI Portable IR Flat Panel Heater provides efficient infrared heating, designed for portability and ease of use in various settings. It operates on 240V, delivering substantial heat output.
  • Operates on 240V for powerful heating.
  • Delivers 14,972 BTU heating capacity.
  • Features a 4.3KW heating output.
  • Designed as a portable unit for flexible placement.
  • Equipped with a roll-about configuration for easy mobility.
  • Utilizes infrared flat panel technology for effective heat transfer.
Catalog No. 50-005-09073 Supplier MSC Supplier No. 55620272
This TPI Portable IR Flat Panel Heater provides efficient infrared heating, designed for portability and ease of use in various settings. It operates on 240V, delivering substantial heat output.
  • Operates on 240V for powerful heating.
  • Delivers 14,972 BTU heating capacity.
  • Features a 4.3KW heating output.
  • Designed as a portable unit for flexible placement.
  • Equipped with a roll-about configuration for easy mobility.
  • Utilizes infrared flat panel technology for effective heat transfer.