Learn More
Aviva Systems Biology Corp TPI1 Antibody - N-terminal region (ARP48144_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48144_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TPI1. Alternate names: MGC88108, TPI, TIMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-59728