Learn More
ABclonal Technology TPI1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A157331000UL
Encompass_Preferred
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate (G3P) and dihydroxy-acetone phosphate (DHAP) in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-06554