Learn More
Medchemexpress LLC HY-12358 5mg Medchemexpress, Tpl2 Kinase Inhibitor 1 CAS:871307-18-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123585MG
Encompass_Preferred
Medchemexpress, HY-12358 5mg Tpl2 Kinase Inhibitor 1 CAS:871307-18-5 Tpl2 Kinase Inhibitor 1 (Compound 1) is a potent and selective Tpl2 (COT kinase, MAP3K8) inhibitor, plays an important role in the regulation of the inflammatory response and the progression of some cancers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9647