Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tpl2 kinase inhibitor 1 | 871307-18-5 | 99.4% | 404.83 g/mol | C21H14ClFN6 | 1MG
SDP

Catalog No. 5000238843
Encompass_Preferred

Tpl2 Kinase Inhibitor 1 is a selective small-molecule inhibitor of Tpl2 (MAP3K8) for use in biochemical and cell-based studies of inflammatory signaling and oncology-related pathways. It inhibits recombinant Tpl2 with an IC50 of 50 nM and reduces LPS-induced TNF-α production in human monocytes and whole blood. The compound is supplied as a yellow to green solid with high chemical purity for research applications.

  • Selective Tpl2 inhibition with IC50 of 50 nM.
  • Cellular activity reducing LPS-induced TNF-α (monocytes IC50 = 0.7 μM; whole blood IC50 = 8.5 μM).
  • High purity suitable for assays (≈99.4%).
  • Molecular formula C21H14ClFN6; molecular weight 404.83 g/mol.
  • Physical form: yellow to green solid.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Applications: biochemical kinase assays and cell-based signaling studies.

Catalog No. 50-002-38843 Supplier Medchemexpress LLC Supplier No. HY123581MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tpl2 Kinase Inhibitor 1 is a selective small-molecule inhibitor of Tpl2 (MAP3K8) for use in biochemical and cell-based studies of inflammatory signaling and oncology-related pathways. It inhibits recombinant Tpl2 with an IC50 of 50 nM and reduces LPS-induced TNF-α production in human monocytes and whole blood. The compound is supplied as a yellow to green solid with high chemical purity for research applications.

  • Selective Tpl2 inhibition with IC50 of 50 nM.
  • Cellular activity reducing LPS-induced TNF-α (monocytes IC50 = 0.7 μM; whole blood IC50 = 8.5 μM).
  • High purity suitable for assays (≈99.4%).
  • Molecular formula C21H14ClFN6; molecular weight 404.83 g/mol.
  • Physical form: yellow to green solid.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Applications: biochemical kinase assays and cell-based signaling studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.