Learn More
Med Vet International TPLO Plate 3.5mm Contour, Lock Screw Left, J0438AL, Each
Catalog No. 5000582644
Encompass_Preferred
The latest generation of TPLO plates involves the use of locking screws. The head of these special screws are threaded to coincide with threads on the inside of the plate hole. JorVet locking screws are made to international standards.
Catalog No. 50-005-82644 Supplier Med Vet International Supplier No. JOR438AL
The latest generation of TPLO plates involves the use of locking screws. The head of these special screws are threaded to coincide with threads on the inside of the plate hole. JorVet locking screws are made to international standards.