Learn More
Aviva Systems Biology Corp TPM1 Antibody - middle region (ARP41443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TPM1. Alternate names: C15orf13, HTM-alpha, TMSA, CMH3, CMD1YApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-57856