Learn More
Aviva Systems Biology Corp TPM2 Antibody - C-terminal region (ARP48224_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48224_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TPM2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TPM2. Alternate names: AMCD1, DA1, TMSB, DA2B, NEM4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53544