Learn More
Aviva Systems Biology Corp TPM3 Antibody - middle region (ARP41141_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41141_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TPM3. Alternate names: FLJ41118, MGC14582, MGC3261, MGC72094, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TPMsk3, TRK, hscp30, TM5, CFTD, hTM5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57735