Learn More
Cayman Chemical TPMPA (hydrate), Purity: ≥95%, 10 mg
SDP
Catalog No. 502936106
Encompass_Preferred
A selective GABAA-ρ1 (ρ1 GABAC) receptor antagonist (Kbs = 2 and 16 μM for GABAA-ρ1 over GABAA-ρ2, respectively); blocks the inhibitory effect of 3-APMPA on EPSCs in rat hippocampal slices (EC50 = 490 μM); increases light sensitivity and the maximal response of rat retinal ganglion cells without altering their dynamic range (100 μM); increases waking from 45 to 62% compared to vehicle control and decreases slow-wave and REM by 11 and 5.4%, respectively, in rats (50 μg, i.c.v.)
Catalog No. 50-293-6106 Supplier Cayman Chemical Supplier No. 2430210 MG
A selective GABAA-ρ1 (ρ1 GABAC) receptor antagonist (Kbs = 2 and 16 μM for GABAA-ρ1 over GABAA-ρ2, respectively); blocks the inhibitory effect of 3-APMPA on EPSCs in rat hippocampal slices (EC50 = 490 μM); increases light sensitivity and the maximal response of rat retinal ganglion cells without altering their dynamic range (100 μM); increases waking from 45 to 62% compared to vehicle control and decreases slow-wave and REM by 11 and 5.4%, respectively, in rats (50 μg, i.c.v.)