Learn More
Medchemexpress LLC HY-100380 5mg Medchemexpress, TPO agonist 1 CAS:1033040-23-1 Purity:>98%
SDP
Catalog No. 501931800
Encompass_Preferred
Medchemexpress, HY-100380 5mg TPO agonist 1 CAS:1033040-23-1 TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1800 Supplier Medchemexpress LLC Supplier No. HY1003805MG
Medchemexpress, HY-100380 5mg TPO agonist 1 CAS:1033040-23-1 TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.