Learn More
Medchemexpress LLC HY-P7088 10ug Medchemexpress, TPO (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP708810UG
Encompass_Preferred
Medchemexpress, HY-P7088 10ug TPO (CHO-expressed), Mouse CAS: Recombinant Mouse Thrombopoietin (CHO-expressed) is a hematopoietic factor that controls platelet production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3106