Learn More
Enzo Life Sciences Thrombopoietin (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX2018080010
Encompass_Preferred
Alternative name: TPO, Megakaryocyte colony stimulating factor, c-MPL ligand, MGDF. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥95% (Reducing and Non-reducing SDS-PAGE). MW: ~18.7kDa. Formulation: Lyophilized. Sterile filtered. Source: Produced in E. coli. Non-glycosylated protein, containing 174 amino acids (which comprise the receptor binding domain). Long Term Storage: -20°C.
Catalog No. 50-200-8011