Learn More
Medchemexpress LLC TPO/Thyroid Peroxida 20ug | 20ug
SDP
Catalog No. 5000187736
Encompass_Preferred
Thyroid Peroxidase (TPO) orchestrates iodination and coupling within thyroglobulin leading to the synthesis of thyroid hormones T(3) and T(4) TPO/Thyroid Peroxidase Protein Mouse (HEK293 His) is the recombinant mouse-derived TPO/Thyroid Peroxidase protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87736 Supplier Medchemexpress LLC Supplier No. HYP7804520UG
Thyroid Peroxidase (TPO) orchestrates iodination and coupling within thyroglobulin leading to the synthesis of thyroid hormones T(3) and T(4) TPO/Thyroid Peroxidase Protein Mouse (HEK293 His) is the recombinant mouse-derived TPO/Thyroid Peroxidase protein expressed by HEK293 with C-His labeled tag