Learn More
Cayman Chemical TPOCTerm RabbIt Monclona 100uL
SDP
Catalog No. 502947814
Encompass_Preferred
An internal standard for the quantification of boceprevir by GC- or LC-MS
Catalog No. 50-294-7814 Supplier Cayman Chemical Supplier No. 32293100 UL
An internal standard for the quantification of boceprevir by GC- or LC-MS