Learn More
Medchemexpress LLC HY-100697 5mg Medchemexpress, TPOP146 CAS:2018300-62-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006975MG
Encompass_Preferred
Medchemexpress, HY-100697 5mg TPOP146 CAS:2018300-62-2 TPOP146 is a selective CBP/P300 benzoxazepine bromodomain inhibitor with Kd values of 134 nM and 5.02 μM for CBP and BRD4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0599