Learn More
Cayman Chemical TPP-1 (hydrochloride), Purity: ≥95%, 25 mg
SDP
Catalog No. 502915648
Encompass_Preferred
A peptide inhibitor of the PD-1-PD-L1 protein-protein interaction; binds to PD-L1 in a cell-free assay (Kd = 94.67 nM); reverses PD-L1-induced decreases in IFN-γ levels in anti-CD3-induced isolated human CD4+ T cells at 20 µM; reduces tumor volume in a mouse xenograft model using HL-60 leukemia cells co-cultured with anti-CD3-activated isolated human CD4+ T cells prior to implantation
Catalog No. 50-291-5648 Supplier Cayman Chemical Supplier No. 3741025 MG
A peptide inhibitor of the PD-1-PD-L1 protein-protein interaction; binds to PD-L1 in a cell-free assay (Kd = 94.67 nM); reverses PD-L1-induced decreases in IFN-γ levels in anti-CD3-induced isolated human CD4+ T cells at 20 µM; reduces tumor volume in a mouse xenograft model using HL-60 leukemia cells co-cultured with anti-CD3-activated isolated human CD4+ T cells prior to implantation