Learn More
Andwin Scientific TPP 300CM TFLASK 18/CS
SDP
Catalog No. NC0245838
Encompass_Preferred
The TPP tissue culture flasks offers a superior growth surface on the flask base for the most varied matrix-dependent tissue cultures. The sides and top inside surfaces are untreated that help to lessen the amount of cell loss during the separation phases, because undesired tissue adhesion outside the growth zone is diminished. The flask features a filtered cap that gives free gas exchange without gas flow alteration. This product is sold in the case of 18.
Catalog No. NC0245838 Supplier Andwin Scientific Supplier No. 90301
The TPP tissue culture flasks offers a superior growth surface on the flask base for the most varied matrix-dependent tissue cultures. The sides and top inside surfaces are untreated that help to lessen the amount of cell loss during the separation phases, because undesired tissue adhesion outside the growth zone is diminished. The flask features a filtered cap that gives free gas exchange without gas flow alteration. This product is sold in the case of 18.