Learn More
Medchemexpress LLC Tpp-CAQK TFA | 99.1% | 906.95 | 1 MG
SDP
Catalog No. 5000425078
Encompass_Preferred
Tpp-CAQK TFA is an engineered mitochondrial compound that can bind to mitochondria, enabling the construction of Mito-Tpp-CAQK TFA with excellent bioactivity. Mito-Tpp-CAQK TFA can be internalized by macrophages, thereby enhancing the phagocytosis of myelin debris, alleviating mitochondrial dysfunction, and reducing proinflammatory profiles, ultimately facilitating tissue repair and functional recovery in mice after spinal cord injury.
  • Can bind to mitochondria
  • Enables construction of Mito-Tpp-CAQK TFA
  • Enhances macrophage phagocytosis of myelin debris
  • Alleviates mitochondrial dysfunction
  • Reduces proinflammatory profiles
  • Facilitates tissue repair and functional recovery in mice after spinal cord injury
  • For research use only
Catalog No. 50-004-25078 Supplier Medchemexpress LLC Supplier No. HYP10707A1MG
Tpp-CAQK TFA is an engineered mitochondrial compound that can bind to mitochondria, enabling the construction of Mito-Tpp-CAQK TFA with excellent bioactivity. Mito-Tpp-CAQK TFA can be internalized by macrophages, thereby enhancing the phagocytosis of myelin debris, alleviating mitochondrial dysfunction, and reducing proinflammatory profiles, ultimately facilitating tissue repair and functional recovery in mice after spinal cord injury.
  • Can bind to mitochondria
  • Enables construction of Mito-Tpp-CAQK TFA
  • Enhances macrophage phagocytosis of myelin debris
  • Alleviates mitochondrial dysfunction
  • Reduces proinflammatory profiles
  • Facilitates tissue repair and functional recovery in mice after spinal cord injury
  • For research use only