Learn More
Medchemexpress LLC Tpp-CAQK TFA | 99.1% | 906.95 | 5 MG
SDP
Catalog No. 5000425205
Encompass_Preferred
Tpp-CAQK TAF can bind to mitochondria, enabling the construction of an engineered mitochondrial compound, Mito-Tpp-CAQK TFA, with excellent bioactivity. Mito-Tpp-CAQK TFA can be internalized by macrophages, thereby enhancing the phagocytosis of myelin debris, alleviating mitochondrial dysfunction, and reducing proinflammatory profiles, ultimately facilitating tissue repair and functional recovery in mice after spinal cord injury.
  • Binds to mitochondria
  • Enables construction of Mito-Tpp-CAQK TFA
  • Enhances phagocytosis of myelin debris
  • Alleviates mitochondrial dysfunction
  • Reduces proinflammatory profiles
  • Facilitates tissue repair and functional recovery in spinal cord injury
Catalog No. 50-004-25205 Supplier Medchemexpress LLC Supplier No. HYP10707A5MG
Tpp-CAQK TAF can bind to mitochondria, enabling the construction of an engineered mitochondrial compound, Mito-Tpp-CAQK TFA, with excellent bioactivity. Mito-Tpp-CAQK TFA can be internalized by macrophages, thereby enhancing the phagocytosis of myelin debris, alleviating mitochondrial dysfunction, and reducing proinflammatory profiles, ultimately facilitating tissue repair and functional recovery in mice after spinal cord injury.
  • Binds to mitochondria
  • Enables construction of Mito-Tpp-CAQK TFA
  • Enhances phagocytosis of myelin debris
  • Alleviates mitochondrial dysfunction
  • Reduces proinflammatory profiles
  • Facilitates tissue repair and functional recovery in spinal cord injury