Learn More
Medchemexpress LLC TPPB | 497259-23-1 | MFCD09037558 | 99.8% | 501.54 g/mol | C27H30F3N3O3 | 10MG
SDP
Catalog No. 5000239390
Encompass_Preferred
TPPB is a cell-permeable benzolactam-derived activator of protein kinase C (PKC) used in biochemical and cell-based research to probe PKC signaling and APP-related pathways. It binds PKC with high affinity (reported Ki ≈ 11.9 nM), and is characterized by molecular formula C27H30F3N3O3 and molecular weight 501.54 g/mol.
  • High-affinity PKC activator (Ki ≈ 11.9 nM).
  • Cell-permeable benzolactam scaffold suitable for cellular assays.
  • Characterized molecular formula C27H30F3N3O3 and molecular weight 501.54 g/mol.
  • High purity confirmed by LCMS (≈99.8%).
  • Recommended storage for the powder: -20°C for long-term stability.
Catalog No. 50-002-39390 Supplier Medchemexpress LLC Supplier No. HY1235910MG
TPPB is a cell-permeable benzolactam-derived activator of protein kinase C (PKC) used in biochemical and cell-based research to probe PKC signaling and APP-related pathways. It binds PKC with high affinity (reported Ki ≈ 11.9 nM), and is characterized by molecular formula C27H30F3N3O3 and molecular weight 501.54 g/mol.
  • High-affinity PKC activator (Ki ≈ 11.9 nM).
  • Cell-permeable benzolactam scaffold suitable for cellular assays.
  • Characterized molecular formula C27H30F3N3O3 and molecular weight 501.54 g/mol.
  • High purity confirmed by LCMS (≈99.8%).
  • Recommended storage for the powder: -20°C for long-term stability.