Learn More
Aviva Systems Biology Corp TPPP3 Antibody - N-terminal region (ARP42457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPPP3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TPPP3. Alternate names: CGI-38, p20, p25gammaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-285-003