Learn More
Sigma Aldrich Fine Chemicals Biosciences TPPR centrifuge tubes capa
Catalog No. 502908887
Encompass
The TPP 50 mL and 15 mL Centrifuge Tubes are compatible with all standard centrifuges and their innovative design focuses on a tight seal excellent transparency clear graduations and highly visible marking areas. The yellow screw cap closure on the TPP Centrifuge Tubes is tight and protected against overturning. The transparency of the tubes is excellent and the white marking areas on the top of the cap and side of the tube are highly visible. On the conical tubes the distinct white graduations are continued to the bottom of the tube to accommodate small sample volumes. On the 500 mL tubes the scale starts at 0.5 mL and on the 15 mL tubes the scale starts at 0.1mL. Temperature Range for Polypropylene Tubes -196C to 121C Polystyrene Tubes -40C to 70C Maximum RCF for Polypropylene Tubes 9400 x g* Polystyrene Tubes 1700 x g*.* Depending on adapter and temperature All Tubes are gamma Irradiated
Catalog No. 50-290-8887 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z707724800EA
The TPP 50 mL and 15 mL Centrifuge Tubes are compatible with all standard centrifuges and their innovative design focuses on a tight seal excellent transparency clear graduations and highly visible marking areas. The yellow screw cap closure on the TPP Centrifuge Tubes is tight and protected against overturning. The transparency of the tubes is excellent and the white marking areas on the top of the cap and side of the tube are highly visible. On the conical tubes the distinct white graduations are continued to the bottom of the tube to accommodate small sample volumes. On the 500 mL tubes the scale starts at 0.5 mL and on the 15 mL tubes the scale starts at 0.1mL. Temperature Range for Polypropylene Tubes -196C to 121C Polystyrene Tubes -40C to 70C Maximum RCF for Polypropylene Tubes 9400 x g* Polystyrene Tubes 1700 x g*.* Depending on adapter and temperature All Tubes are gamma Irradiated